Capillary columns, Rtx®-CLPesticides (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42102 42301 42302 43101 43301 11120 11123 11126 11136 11139 11140 11321 11323 11324 11326 11337 11340 11123-600 11139-600
REST42102EA 5400 DKK
REST42102 REST42301 REST42302 REST43101 REST43301 REST11120 REST11123 REST11126 REST11136 REST11139 REST11140 REST11321 REST11323 REST11324 REST11326 REST11337 REST11340 REST11123-600 REST11139-600
Capillary columns, Rtx®-CLPesticides (fused silica)
Kromatografikolonner
Application specific columns for organochlorine pesticides and herbicides. Low bleed ideal for GC-ECD or GC-MS analyses.
Proprietary Crossbond® phases
Temperature range: Stable to 340 °C
Analyse EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1 and 552.2 compounds without time-consuming column change.