Solid phase extraction cartridges for pesticide residue cleanup
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26127 26128 26129 26193 26194
REST26127EA 2910 DKK
REST26127 REST26128 REST26129 REST26193 REST26194
Solid phase extraction cartridges for pesticide residue cleanup
Kromatografikolonner
Pesticide residue cleanup SPE cartridges are convenient with multiple adsorbent beds in a single cartridge. It is used in multiresidue pesticide analysis to remove matrix interferences and also exhibits good cleanup of dietary supplement extracts.