Capillary columns, Rtx®-502.2 (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40914 40915 10908 10909 10910 10915 10916 10919 10920 10921 10986
REST40914EA 4790 DKK
REST40914 REST40915 REST10908 REST10909 REST10910 REST10915 REST10916 REST10919 REST10920 REST10921 REST10986
Capillary columns, Rtx®-502.2 (fused silica)
Kromatografikolonner
Application-specific columns with unique selectivity for volatile organic pollutants. The Rtx®-502.2 column is cited in U.S. EPA method 502.2 and in many gasoline range organics (GRO) methods for monitoring underground storage tanks. For separation of trihalomethanes, ideal polarity for light hydrocarbons and aromatics.
Proprietary Crossbond® diphenyl/dimethyl polysiloxane phase.
Temperature range: Stable to 270 °C