Capillary columns, Stabilwax®-DA (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11005 11007 11008 11009 11020 11021 11022 11023 11024 11025 11026 11027 11028 11035 11036 11037 11038 11039 11040 11041 11042 11043 11051 11052 11055 11057 11058 11062 11065 11068
REST11005EA 3630 DKK
REST11005 REST11007 REST11008 REST11009 REST11020 REST11021 REST11022 REST11023 REST11024 REST11025 REST11026 REST11027 REST11028 REST11035 REST11036 REST11037 REST11038 REST11039 REST11040 REST11041 REST11042 REST11043 REST11051 REST11052 REST11055 REST11057 REST11058 REST11062 REST11065 REST11068
Capillary columns, Stabilwax®-DA (fused silica)
Kromatografikolonner
Application specific columns for free (underivatized) acids and some inorganic acids. Resistant to oxidative damage.
Polar phase; Crossbond® acid deactivated Carbowax® polyethylene glycol for acidic compounds
Stabilwax®-DA bonded polyethylene glycol has an acidic functionality incorporated into the polymer structure. This permits analysis of acidic compounds without derivatisation, significantly reduces adsorption of acids and increases sample capacity for volatile free acids. These columns give better peak shapes for high molecular weight acids.
Temperature range: 40 to 260 °C
Similar phases: HP-FFAP, DB-FFAP, CP-WAX 58 FFAP CB, NUKOL, ZB-FFAP. Equivalent to USP G25, G35 phases.