HPLC columns, Allure® C18 (USP L1)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9164512 9164515 9164531 9164551 9164552 9164555 9164562 9164565 9164572 9164575
REST9164512EA 0 DKK
REST9164512 REST9164515 REST9164531 REST9164551 REST9164552 REST9164555 REST9164562 REST9164565 REST9164572 REST9164575
HPLC columns, Allure® C18 (USP L1)
Kromatografikolonner
Allure® C18 columns are designed using a unique bonding chemistry that offers resistance to hydrolysis and superior batch to batch reproducibility. The high carbon load phase exhibits maximum retention characteristics, making it ideal for liquid chromatography/mass spectrometry (LC/MS) applications. The phase is bonded onto a highly base deactivated type B silica, resulting in improved peak symmetry, even for basic analytes. The column can be used to analyse compounds, steroids, antibiotics and explosives.