Transparent bath tanks, polycarbonate, for CORIO C / CD
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9901305 9901309 9901319 9901327
461-0900EA 1660 DKK
461-0900 461-0901 461-0902 461-0903
Transparent bath tanks, polycarbonate, for CORIO C / CD
Bade Vandbade, uden omrøring
Transparent PC bath tanks in different sizes. For use with all CORIO C and CD circulators.
Temperature range: +20...+100 °C