Industrial balances, Ranger Count 4000 series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30231102 30231103 30231104 30231105 30237020 30237021 30237022 30237023
611-3411EA 6070 DKK
611-3411 611-3402 611-3412 611-3413 611-3419 611-3420 611-3421 611-3422
Industrial balances, Ranger Count 4000 series
Vægte
A multipurpose compact counting scale for a wide variety of tough industrial counting tasks. Applications include weighing, parts counting, check counting and accumulation functions.
  • Rugged die-cast metal housing and slip resistand rubber feet provde protection and stability
  • Maximise throughput without sacrificing accuracy
  • Three separate backlit displays for count, weight and piece weight
  • Mains supply with internal rechargeable battery (lifetime up to 210 hours) for portable use
  • Internal memory with a 30 item library function for data including average piece weights, check counting or check weighing, parameters, tare weights and accumulated weights or counts
  • Stabilisation within 1 second
Certificeringer: Meets Class III requirements of OIML R76 and EN 45501
Bestillingsoplysninger: More connectivity options available including optional ethernet, second RS232 or USB and optional second platform. Approved versions are also available. Please enquire for more details.