Industrial bench scales, Ranger 4000 series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30236943 30231099 30231100 30231101 30237016 30237017 30237018 30237019
611-3414EA 6030 DKK
611-3414 611-3408 611-3409 611-3410 611-3415 611-3416 611-3417 611-3418
Industrial bench scales, Ranger 4000 series
Vægte
Multipurpose, compact scale for producing precise results rapidly. Application modes include weighing, percent weighing, parts counting, accumulation and dynamic weighing and display hold
  • Bright white LED display, suitable for dim light conditions
  • Portable (net weight 5,9 kg) with rugged die-cast metal housing and rechargeable battery
  • Simple keyboard; five key operation and intuitive SmarText™ software
  • Stabilisation within 1 second
  • Checkweighing LEDs and audible indicators
Certificeringer: Meets Class III requirements of OIML R76 and EN 45501
Bestillingsoplysninger: More connectivity options available including optional ethernet, second RS232 or USB and optional second platform. Approved versions are also available. Please enquire for more details.