Capillary columns, Rxi®-5ms (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13423 I13438 13420 13421 13439 13424 13455 13402 13481 13426 13442 13409 13453
REST13423EA 6050 DKK
REST13423 JJBEI13438 REST13420 REST13421 REST13439 REST13424 REST13455 REST13402 REST13481 REST13426 REST13442 REST13409 REST13453
Capillary columns, Rxi®-5ms (fused silica)
Kromatografikolonner
General purpose columns for semivolatiles, phenols, amines, residual solvents, drugs of abuse, pesticides, PCB congeners (e.g. Aroclor mixes), solvent impurities.
Low polarity phase; Crossbond® 100% diphenyl dimethyl polysiloxane
Inert column, tested and guaranteed for ultra-low bleed, improved signal-to-noise ratio for better sensitivity and mass spectral integrity.
Temperature range: −60 to 330/350 °C
Similar phases: HP-1 ms, HP-1 msUI, DB-1 ms, DB-1 msUI, Ultra-1, VF-1 ms, ZB-1, ZB-1 ms.
Equivalent to USP G1, G2 and G38 phases.