VWR® EML 200, testsigte-rystere
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
510-2043EA 17420 DKK
510-2043 510-2045 510-2046 510-2048 510-2037 510-2039 510-2049 510-2051 510-2040 510-2042 HAVE205323041 HAVE205323058 HAVE205323027 HAVE205322990 HAVE205323010
VWR® EML 200, testsigte-rystere
Rystere Sigterystere
The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a cast aluminium housing and improved clamping system.
  • Digital timer: 0 to 99 minutes or constant operation
  • All shakers have a cover with inspection glass which allows samples to be viewed during sieving
EML 200 Pure units are ideal for standard analysis:
Feature two given regulated amplitudes to select from: Fine or coarse
Operation in set intervals of 10 seconds
Classic 'eco' clamping system
EML 200 Premium units are suitable for users requiring precise and reproducible sieve analyses:
Extra quiet electromagnetic drive
Freely selectable amplitudes up to 3 mm
Extra large, easy to use display and simple logical operation
During sieving the remaining sieving time is shown on the display
Up to 49 different sieving programs can be stored in the memory
Option of Classic or HAVER TwinNut® quick release clamping system
EML 200 Premium Remote models also offer wet sieving, features as Premium units.
Integrated full cone nozzle
Separate controller which can be wall mounted
Bestillingsoplysninger: Acceptance test certificate 3.1 according to DIN EN 10204 and IQ/OQ documentation for are available as optional accessories for all models, for details please contact your local Avantor sales office.