Temperature control jackets for viscosity cups
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VF2067 VF2068
769-1131EA 4060 DKK
769-1131 SIMEVF2068
Temperature control jackets for viscosity cups
Styreenheder Temperaturkontroller
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
  • Adjustable feet for easy leveling
  • Built-in spirit level to set jacket exactly horizontal
  • Glass plate to prevent dripping before measurement
  • Elbow-type quick release couplings
  • Titanium anodised aluminium
Leveringsinformation: Supplied with jacket with built-in spirit level, glass plate and tube connection.