Cut resistant gloves, HyFlex® 11-735
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-735/6 11-735/7 11-735/9 11-735/8 11-735/11 11-735/10
112-4583EA 1640 DKK
112-4583 112-4584 112-4586 112-4585 112-4582 112-4581
Cut resistant gloves, HyFlex® 11-735
Handsker
This seamless plaited liner with INTERCEPT™ technology yarn and dipped PU coating on palm and fingertips provides high levels of cut protection.
  • INTERCEPT™ yarn offers excellent cut protection and increased comfort and dexterity
  • Palm coating design improves grip, extends glove life in abrasive conditions and allows the back of the hand to breath
  • Silicone-free PU coating is more process friendly in use when metal parts are painted
  • Grey coating hides dirt in environments with dirt exposure
Ideal for the assembly of metal parts, components and white goods, for general maintenance and for metal fabrication.
Certificeringer: EN 388:2016 (4X43C), Cat. II