Equivalence C18, HPLC Columns
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EQV-5C18-2546EA 3350 DKK
EQV-5C18-2546 EQV-3C18-0546 EQV-3C18-1046 EQV-3C18-1546 EQV-5C18-1546 EQV-3C18-GD EQV-5C18-GD EQV-3C18-1503 EQV-3C18-0502 EQV-3C18-1002 EQV-3C18-7546 EQV-3C18-1246 EQV-5C18-0546 EQV-5C18-7546 EQV-5C18-1046 EQV-5C18-1246 EQV-5C18-1504 EQV-5C18-2504 EQV-3C18-1203 EQV-5C18-1003 EQV-5C18-1203 EQV-3C18-1502 EQV-5C18-1002 EQV-3C18-GD2 EQV-3C18-0346
Equivalence C18, HPLC Columns
Kromatografikolonner
Stainless steel Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
  • Excellent efficiency and reproducibility
  • Available in 3 or 5 µm particle sizes
Maximum recommended pH: 2,0 to 8,0