Gas detection tubes for toluene
Leverandør: Dräger
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
118-0313EA 1120 DKK
118-0313 118-0316 118-0315
Gas detection tubes for toluene
Gasslanger
To measure the 'spot concentrations' of toluene for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, measurement of personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.