VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0636EA 669 DKK
525-0636 525-0637
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Rør Centrifugerør
Made of high quality, USP class 6 gamma-resistent clear polypropylene with HDPE screw cap.
  • Longer length screw caps with sealing ring prevent leakage
  • Easy-to-read black graduations are accurate to ±2%, with 1 ml increments (15 ml) or 2,5 ml increments (50 ml)
  • Large unerasable frosted white writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Autoclavable at 121 °C and freezable to −80 °C
  • Leakproof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Disposable conical-bottom centrifuge tubes with smooth inner walls for easy filling and sample preparation. A white printed writing area on the side allows for permanent coding. Tubes are sterilised by irradiation. RNase-free, DNase-free and non-pyrogenic.
Certificeringer: TSE/BSE-free Certification.
Leveringsinformation: Bagged tubes are packaged in durable and resealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability. All tubes are packaged with caps attached and sterilised with irradiation.