HPLC columns, Hibar®, LiChrospher® 100 RP-8e
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00423.7091 1.00423.7092 1.50012.7091 1.50013.7091 1.50182.7091 1.50182.7092 1.50183.7091 1.50183.7092
1.00423.7091EA 7340 DKK
1.00423.7091 1.00423.7092 1.50012.7091 1.50013.7091 1.50182.7091 1.50182.7092 1.50183.7091 1.50183.7092
HPLC columns, Hibar®, LiChrospher® 100 RP-8e
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.