HPLC columns, Hibar®, LiChrospher® 100 DIOL
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.00424.7075 1.50013.7075 1.50183.7075 1.50012.7075
1.00424.7075EA 7340 DKK
1.00424.7075 1.50013.7075 1.50183.7075 1.50012.7075
HPLC columns, Hibar®, LiChrospher® 100 DIOL
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.