HPLC columns, LiChroCART®, Purospher® RP-18e
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50171.7130 1.50178.7130 1.50179.7130 1.50195.7130 1.50201.7130 1.50228.7130 1.50229.7130 1.50233.7130 1.50234.7130 1.50236.7130 1.50302.7130 1.51431.7130 1.51442.7130 1.51443.7130 1.51444.7130 1.51448.7130 1.50175.7130 1.50177.7130 1.50173.7130 1.50170.7130 1.50174.7130
1.50171.7130EA 0 DKK
1.50171.7130 1.50178.7130 1.50179.7130 1.50195.7130 1.50201.7130 1.50228.7130 1.50229.7130 1.50233.7130 1.50234.7130 1.50236.7130 1.50302.7130 1.51431.7130 1.51442.7130 1.51443.7130 1.51444.7130 1.51448.7130 1.50175.7130 1.50177.7130 1.50173.7130 1.50170.7130 1.50174.7130
HPLC columns, LiChroCART®, Purospher® RP-18e
Kromatografikolonner
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.