VWR®, Test Sieves, 305×60 mm, with Square Holes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
510-2929EA 2100 DKK
510-2929 510-2930 510-2931 510-2932 510-2933 510-2934 510-2935 510-2936 510-2937 510-2938 510-3300 510-3301 510-3302 510-3303 510-3304 510-3305 510-3306 510-3307 510-3308 510-3309 510-3310 510-3311 510-3312 510-3313 510-3314 510-3315 510-3316 510-3317 510-3318 510-3319 510-3320 510-3321 510-3322 510-3323 510-3324 510-3325 510-3326 510-3327 510-3328 510-3329 510-3330
VWR®, Test Sieves, 305×60 mm, with Square Holes
Sigter Testsigter
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Leveringsinformation: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.