Glass microfibre filters without binder, grade 934-AH, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1827-024 1827-032 1827-042 1827-047 1827-055 1827-070 1827-090 1827-110 1827-125 1827-866 1827-025 1827-021 1827-082 1827-035 1827-808 1827-889 1827-028 1827-030 1827-037 1827-085 1827-105 1827-150 1827-185 1827-240 1827-320 1827-957
516-2704EA 318 DKK
516-2704 512-0428 513-5275 516-2711 516-2713 516-2715 516-2716 516-2717 512-0429 513-5276 513-5274 513-5273 512-0668 513-0109 WHAT1827-808 513-0111 WHAT1827-028 513-0108 513-0110 512-0669 516-5107 516-5108 516-5109 516-5112 516-5114 512-0670
Glass microfibre filters without binder, grade 934-AH, Whatman™
Filtre Glasfiberfiltre
These borosilicate glass filters are specified for water pollution monitoring, cell harvesting, liquid scintillation counting and air pollution monitoring. They are often used in total suspended solids analysis.
  • Smooth surface for efficient precipitate recovery
  • High loading capacity
  • Binder free
Total suspended solids analysis (previously called non-filterable residue) requires high loading capacity glass microfiber filters, such as grade 934-AH.