VWR®, Cryoboxes with Dividers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
479-1437EA 42.3 DKK
479-1437 479-1438 479-1439 479-1440 479-1441 479-1442
VWR®, Cryoboxes with Dividers
Bokse Æsker til Kryorør
Boxes, made of cardboard, white. For storage of cryovials at low temperatures.
  • Resistant to temperatures down to –140 °C
  • Standard waterproof coating