VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0633EA 1060 DKK
525-0633 525-0635 525-0634
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Rør Centrifugerør
Manufactured of high-quality USP Class 6 gamma resistent clear polypropylene with HDPE screw cap.
  • Longer length screw caps
  • Easy-to-read black graduations are accurate to ±2%
  • Large unerasable frosting white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Autoclavable at 121 °C and freezable to –80 °C
  • Leak-Proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
These disposable freestanding tubes feature black printed graduations and smooth inner walls for easy filling and sample preparation. A white printed writing area on the side allows for permanent coding. All tubes are RNase-free, DNase-free, and non-pyrogenic.
Certificeringer: TSE/BSE-free Certification.
Leveringsinformation: Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Inner packaging is individually labelled for lot-to-lot traceability.