Sampling pouches, 500 series Kapak®
Leverandør: ProAmpac
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
506B-500 504-24 505-24 502-24 503-24 500-24 500B-1M 502B-1M 503B-1M 504B-1M 505B-1M 506-24 508-24 508B-500
300-0317EA 8730 DKK
300-0317 300-0312 300-0314 300-0308 300-0310 300-0306 300-0307 300-0309 300-0311 300-0313 300-0315 300-0316 300-0318 300-0319
Sampling pouches, 500 series Kapak®
Poser Prøveposer
114 micron high barrier, crystal clear, three side seal, heat sealable polyester pouches.
  • Pouches are microwaveable, boilable and freezable
  • High barrier leak proof pouches help eliminate cross contamination of dangerous substances
  • With 9 mm seals
  • Infrared and gamma ray resistant
  • Once sealed, these pouches can also be surface sterilised using VHP
These pouches can be used for: tissue and blood samples, formalin samples, allografts, sample retains, overwrap for medical trays and kits, medical device sterilisation - gamma irradiation, introducing supplies into sterility testing chambers - surface sterilise secondary packaging using VHP, disposal of contaminated waste, disposal of sharp objects, overwrap for evidence kits and evidence/narcotics storage.
Typical applications: Medical, industrial, food preservation, veterinarian, health and beauty, agriculture.