Safety wash bottles, narrow neck, right-to-understand, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2428-0502 2428-0503 2428-0504 2428-0505 2428-0506 2428-0507
215-3829EA 517 DKK
215-3829 215-3830 215-3831 215-3832 215-3833 215-3834
Safety wash bottles, narrow neck, right-to-understand, Nalgene®
Flasker Sprøjteflasker
LDPE, translucent.
  • Self-venting wash bottles eliminate risk of bottles self-dispensing
  • Good chemical resistance
  • Good temperature resistance
Prevent chemical accidents in the lab with Right-to-Understand wash bottles. Bottles are pre-labeled with chemical identification and safe handling information following the Globally Harmonized System (GHS) specifications as applicable to secondary laboratory containers. Wash bottles are ready to use when you transfer common laboratory chemicals from manufacturers’ containers.
These cylindrical bottles are notMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering NAR Certificate of Retirement