Glass microfibre filters without binder, grade MN GF-6
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4160025 4160037 4160045 416005 416007 416009 416011 416012 416015 416018 416024 416027
MANA4160025EA 525 DKK
MANA4160025 MANA4160037 MANA4160045 MANA416005 MANA416007 MANA416009 MANA416011 MANA416012 MANA416015 MANA416018 MANA416024 MANA416027
Glass microfibre filters without binder, grade MN GF-6
Filtre Glasfiberfiltre
MN GF-6 is a binder-free glass fibre filter paper with medium retention and flow rate for general laboratory filtration.
  • All-round glass fibre filter
  • Medium flow and retention capacity