Glass microfibre filters without binder, grade MN GF-5
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4150025 4150037 4150045 415005 415007 415009 415011 415012 415015 415018 415024 415027
MANA4150025EA 1210 DKK
MANA4150025 MANA4150037 MANA4150045 MANA415005 MANA415007 MANA415009 MANA415011 MANA415012 MANA415015 MANA415018 MANA415024 MANA415027
Glass microfibre filters without binder, grade MN GF-5
Filtre Glasfiberfiltre
MN GF-5 is a binder-free glass fibre filter paper with excellent retention of fine particles. It is often used in combination with membranes to protect the membrane from clogging.
  • Suitable for membrane protection