Glass microfiber filters without binder, grade MN GF-3
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4130045 413009 413011 4130025 4130037 413005 413007 413012 413015 413018 413024 413027
MANA4130045EA 718 DKK
MANA4130045 MANA413009 MANA413011 MANA4130025 MANA4130037 MANA413005 MANA413007 MANA413012 MANA413015 MANA413018 MANA413024 MANA413027
Glass microfiber filters without binder, grade MN GF-3
Filtre Glasfiberfiltre
MN GF-3 is slightly more retentive than MN GF-1. This binder-free glass fiber filter paper is used for the collection of suspended solids in potable water as well as natural and industrial waste water in many parts of the world.
  • For suspened solids determination
  • Binder-free
  • For waste water analysis
Bestil nu
Specifikationer
Environmentally Preferable
Vægt 50 g/m²
Tykkelse 0,28 mm
Partikelretention 0,6 μm
Filtreringshastighed 25 s (air)
Max. temperature 500 °C