Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 3 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MDKE-3-2546UEA 14480 DKK
MDKE-3-2546U MDKE-3-2503U MDKE-3-2502U MDKE-3-1546U MDKE-3-1503U MDKE-3-1502U MDKE-3-1046U MDKE-3-1003U MDKE-3-1002U MDKE-3-0546U MDKE-3-0503U MDKE-3-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 3 µm
Kromatografikolonner
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 and 5 µm) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Maximum recommended pH: 2,0 to 8,0 (1,5 to 11,5 for SuperC18).
Max. pressure: 1000 bar.
Leveringsinformation: Avantor® ACE® Extended Method Development Kits contain Avantor® ACE® SuperC18™, Avantor® ACE® C18-Amide and Avantor® ACE® CN-ES stainless steel columns.