Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 2 µm
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MDKA-2-1002UEA 8820 DKK
MDKA-2-1002U MDKA-2-1546U MDKA-2-1503U MDKA-2-1502U MDKA-2-1046U MDKA-2-1003U MDKA-2-0546U MDKA-2-0503U MDKA-2-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 2 µm
Kromatografikolonner
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Maximum recommended pH: 2,0 to 8,0
Max. pressure: 1000 bar
Leveringsinformation: Avantor® ACE® Advanced Method Development Kits contain Avantor® ACE® C18, Avantor® ACE® C18-AR and Avantor® ACE® C18-PFP stainless steel columns.