Operating supplies
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9910075900 190025400
VARI9910075900EA 1480 DKK
VARI9910075900 VARI190025400
Operating supplies
Spektrofotometer og fotometer Flammefotometre
The operating supplies set includes two 500 ml constant pressure bottle, a diluent bottle (1 L), a three-way SIPS tee piece assembly for SIPS-20, six SIPS pump tubings, a SIPS kit and ten pack SIPS pump bands.