Refrigerated/heating circulators, CORIO™ CD series
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9012703 9012701 9012702 9012704 9012705 9012707 9012708 9012713.N1 9012713.S1 9012714.N1 9012714.S1 9012716.N1 9012715.N1 9012717.N1 9012701.N1 9012728.N1
461-0885EA 35650 DKK
461-0885 461-0883 461-0884 461-0886 461-0964 461-0998 461-0999 461-1455 JULA9012713.S1 461-1456 JULA9012714.S1 461-1457 JULA9012715.N1 JULA9012717.N1 JULA9012701.N1 JULA9012728.N1
Refrigerated/heating circulators, CORIO™ CD series
Cirkulatorsystemer Åbne cirkulatorsystemer Termostatbade til opvarmning/køling
These powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO™ CD controller is an advanced unit with professional technology for demanding applications and can be used for temperature control of internal or external applications. The jet nozzle allows continuous adjustment of the pump stream in the system. The insulated stainless steel baths are durable and have an integrated drain tap. These heating/cooling circulators are ideal for temperature control of samples in a circulator bath or temperature control of an external application. For example: Measuring cells, refractometers, polarimeters, photometers, viscometers, fermenters, electrophoresis chambers, chromatography columns, rotary evaporators and rheometers.
  • Bright, white, easy to read display
  • Very quiet and easy operation
  • Have a USB interface and pump that can change between internal and external circulation via a lever located directly below the display
  • Integrated cooling coil for counter-cooling gives more usable space in the bath tank
  • Refrigeration unit without side vents and removable ventilation grid
  • Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
CD: 15 L/min; 0,35 bar
Certificeringer: Classification III according to DIN 12876-1
IP protection class according to DIN EN 60529: IP 21
Bestillingsoplysninger: An extensive range of accessories mean that the circulators can be modularly and individually adapted to applications.
Leveringsinformation: Supplied with bath cover and two each barbed fitting for tubing 8 and 12 mm inner dia. (pump connections M16×1 male).
Advarsel: Note: Cooling capacity at +20; 0; −10; −20; −30 and −40 °C