Wafer carrier handles for 150 mm wafer handling
Leverandør: ENTEGRIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A72-60-04-0215 A72-60-03-0215
216-1634EA 0 DKK
216-1634 ENTEA72-60-03-0215
Wafer carrier handles for 150 mm wafer handling
Produkter til håndtering af wafers Wafer Carriers
Squeeze style
Attaches to the notches on both end walls of carrier
Overall size: 163×140×183 mm
Compatible with all 'M' style carriers
Slingshot style
Slides onto the top flanges of the carrier
Overall size: 15×192×237 mm
Compatible with all 182 series and 194 series wafer carriers