Floor scales, Defender® 3000, DF series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
83997584 83997585 83997593 83997594 83997595 83997596 83997597 83997598
611-3525EA 12050 DKK
611-3525 611-3526 611-3527 611-3528 611-3529 611-3530 611-3531 611-3532
Floor scales, Defender® 3000, DF series
Vægte
Painted carbon steel platform with 5 mm thick, fixed, top plate with four R60 alloy load cells with IP 67 rating. Easy to use balance with 5 weighing units and basic parts counting functionality.
  • T32MC indicator with ABS plastic face with backlit LCD display
  • IP 65 stainless steel junction box with top access
  • Pit frames and ramps are available to fit your installation needs