Photometric test kits, COD VARIO tube tests
Leverandør: Lovibond
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2420721 2420720 2420722 2420710 2420711 2420712 2420725 2420726 2420727
705-0927EA 1020 DKK
705-0927 705-0926 705-0928 LOVI2420710 LOVI2420711 LOVI2420712 705-0929 705-0930 705-0931
Photometric test kits, COD VARIO tube tests
Miljø test kit
ST-COD ( sealed tube-chemical oxygen demand) tests are ready for use.
  • Suppression of chloride interference; up to 1000 mg/L (LR and MR);up to 10000 mg/L (HR)
  • Mercury-free tube tests, in absence of chloride interference
  • High range (HR) tubes (0 to 15000 mg/L O₂) are suitable for waste water testing
* without chloride removal.