Garment racks, upright and wall mounted
Leverandør: Metro International
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GRU2436S GRU2448S GRU2460S GRU2472S
METOGRU2436SEA 22380 DKK
METOGRU2436S METOGRU2448S METOGRU2460S METOGRU2472S
Garment racks, upright and wall mounted
Møbler Tøjstativer
Racks are designed to provide efficient storage of cleanroom garments. The hangers are spaced at 75 mm intervals.
  • Stainless steel 304 with electropolished finish
  • Open-wire design promotes laminar air flow
  • Adjustable stainless steel levelling feet provides floor mount or free standing option
  • Unique hanger capture mechanism and hanger design provide theft deterrence
ESD classification: non-conductive.