Adapter fittings, Cynergy®
Leverandør: ENTEGRIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SA4-6FN-1 SA8-8FN-1 SA16-16FN-1 SA12-12FN-1 SA16-12FN-1 SA24-16FN-1 SA4-4FN-1
ENTESA4-6FN-1EA 0 DKK
ENTESA4-6FN-1 ENTESA8-8FN-1 ENTESA16-16FN-1 ENTESA12-12FN-1 ENTESA16-12FN-1 ENTESA24-16FN-1 ENTESA4-4FN-1
Adapter fittings, Cynergy®
Fittings og stik
Cynergy® Clamp-to-Flaretek® adapter. These adapters show standard Entegris® fitting port connections used in conjunction with Cynergy® clamp fittings.
Using reliable Flaretek® fitting technology, these adapters provide a clean, single-fitting transition to other Entegris® critical fluid management products used in ambient temperature utility line and process applications. Maximum temperature and pressure ratings are size dependent for the Flaretek® fitting connection.