Chemical resistant gloves, AlphaTec® 58-128
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
58-128/9 58-128/10 58-128/11 58-128/7 58-128/8
112-4393EA 1040 DKK
112-4393 112-4389 112-4390 112-4391 112-4392
Chemical resistant gloves, AlphaTec® 58-128
Handsker
These nylon gloves with thin nitrile coating provide protection against chemicals, oils and other irritant fluids, such as biocides, pesticides, fungicides, paints, inks and dyes.
  • Flexible open weave knit liner and stretchy nitrile formulation coating for better dexterity, tactility and comfort
  • 'Thin Nitrile Technology' formulation: Provides optimal flexibility and enables the hand to move naturally without constriction
  • Ansell Grip Technology™ offers more precise handling of small oily or wet parts
  • Extended cuff provides forearm splash protection
Ideal for agriculture applications, chemical handlings, light assembly in chemical environments, refining, metal working with corrosives, packaging and filling.
Certificeringer: EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A); Cat. III. Food contact approved.