Chemical resistant gloves, AlphaTec® 38-612
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
38-612/10 38-612/9
112-4386EA 1780 DKK
112-4386 112-4388
Chemical resistant gloves, AlphaTec® 38-612
Handsker
These gloves made from Viton/Butyl provide excellent resistance to the most aggressive chemicals, such as aliphatic, halogenated and aromatic hydrocarbons as well as concentrated mineral acids.
  • Very flexible and provide excellent dexterity
  • Natural, curved ergonomic shape and soft feel
  • Easy donning and good grip
Ideal for use in chemical plants, chemical transportation and processing, oil refining and petrochemicals, disaster response and HazMat work.
Certificeringer: EN ISO 374-1:2016 Type A (BCFLPST), EN ISO 374-5:2016, EN 388:2016 (1010X); Cat. III