Gas cylinder trolleys for 10 liter gas cylinders
Leverandør: DUPERTHAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7.00.617 7.00.618
139-0607EA 4070 DKK
139-0607 139-0608
Gas cylinder trolleys for 10 liter gas cylinders
Vogne
For the safe transport of small gas cylinders. Steel tube construction in slate grey with solid rubber tires. Perfect for equipping gas cylinder cabinets and depots. Colour: RAL 7015, coarse structure.
  • Easy handling, sturdy design and optimal radius of movement
  • With securing belt, tool box including suitable wrench for gas cylinder caps