Circular spacers
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
51001135 51001137 51001140 51001141
PERC51001135EA 2010 DKK
PERC51001135 PERC51001137 PERC51001140 PERC51001141
Circular spacers
Afstandsstykker
These circular spacers are designed to be used for the analysis of less viscous samples.
  • Available in 0,025 to 0,500 mm thickness
Since the demountable cells uses the circular windows, circular spacers should be used.