ESD safety ankle boots, lace-up, Dimension Pro, Horizon Mid
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
767821/39 767821/36 767821/37 767821/38 767821/40 767821/41 767821/42 767821/43 767821/44 767821/45 767821/46 767821/47 767821/48
114-7827EA 701 DKK
114-7827 114-7824 114-7825 114-7826 114-7828 114-7829 114-7830 114-7831 114-7832 114-7833 114-7834 114-7835 114-7836
ESD safety ankle boots, lace-up, Dimension Pro, Horizon Mid
Sko og støvler
Leather-free anti-slip ESD safety shoes made from hydrophobised microfibre and hydrophobised textile Cordura® (tear and abrasion resistant, dirt and water repellent) upper, a full padded bellows tongue and a slip resistant dual-density TPU/PU sole Dimension Pro.
  • Climate regulating, breathable textile lining
  • Semi-orthopaedic, anatomically shaped insole supports the natural heel-to-toe movement of the foot
  • Replaceable insole; orthopaedic insoles possible
  • Soft fleece ESD insole
  • Sole prevents twisting and slipping
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/silver
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.