ESD safety ankle boots, lace-up, Trainers, Sander Pro GTX
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
768571/38 768571/39 768571/40 768571/41 768571/42 768571/43 768571/44 768571/45 768571/46 768571/47
114-6256EA 873 DKK
114-6256 114-6257 114-6258 114-6259 114-6260 114-6261 114-6262 114-6263 114-6264 114-6265
ESD safety ankle boots, lace-up, Trainers, Sander Pro GTX
Sko og støvler
100% metal- and leather-free, lightweight ESD safety ankle boots made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Gore-Tex® climate membrane
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20345; S3.