Bottle-top dispensers, Dispensette® S Organic
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4630130 4630131 4630140 4630141 4630150 4630151 4630160 4630161 4630170 4630171 4630230 4630231 4630240 4630241 4630290 4630291 4630330 4630331 4630340 4630341 4630350 4630351 4630360 4630361
613-5330EA 3128 DKK
613-5330 613-5331 613-5332 613-5333 613-5334 613-5335 613-5336 613-5337 613-5338 613-5339 613-5340 613-5341 613-5342 613-5343 613-5344 613-5345 613-5346 613-5347 613-5348 613-5349 613-5350 613-5351 613-5352 613-5353
Bottle-top dispensers, Dispensette® S Organic
Flasketopdispenser
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: fixed (fixed-volume for standard applications; simple calibration adjustment with supplied tool), analog (analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool) and digital
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • DE-M marking
  • Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S Organic is ideal for dispensing organic solvents such as chlorinated and fluorinated hydrocarbons like trichlorotrifluoroethane and dichloromethane, or acids like concentrated HCl and HNO₃ (except for HF), as well as for trifluoroacetic acid (TFA), tetrahydrofurane (THF), and peroxides. For dispensing hydrofluoric acid (HF), we recommend the use of the Dispensette® S Trace Analysis bottle-top dispenser with platinum-iridium valve spring. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, tantalum, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: yellow.
* Special fixed volumes: 0,5 to 100 ml (please state when ordering)!
Bestillingsoplysninger: Dispensette® S Organic supplied with DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and PP adapters.