Kemikaliebestandige handsker, UltraNeo 450
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34450318 34450319 34450000 34450317
112-0688EA 82.1 DKK
112-0688 112-0689 112-0685 112-0687
Kemikaliebestandige handsker, UltraNeo 450
Handsker
Neoprene/natural latex gloves with cotton flock lining.
  • Extremely supple with freedom of movement
  • Provides standard chemical protection
  • Added comfort thanks to the cotton flock lining
  • Embossed textures for good grip on slippery objects
  • Long cuff for forearm protection
Ideal for use in the automotive/mechanical industry, for battery manufacturing, the preparation of industrial glues and for chemical handlings.
Certificeringer: EN 388:2016 (2121X), EN ISO 374-1 Type A (ALMNST), EN ISO 374-5; Cat. III.