Membrane filtration set
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LCW904 LCW916
HACHLCW904EA 1620 DKK
HACHLCW904 HACHLCW916
Membrane filtration set
Filtre Sprøjtefiltre
This membrane filtration set can be used for filtration of turbid samples for the determination of dissolved ingredients.
Leveringsinformation: The set contains 50 syringe filters and one disposable 10 ml syringe.