Production and laboratory chairs, with permanent contact backrest, Neon 3
Leverandør: bimos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2000-3278-525 2000-3279-525 2000-3280-525 2001-3278-525 2001-3279-525 2001-3280-525 MG01-3278-525 MG01-3279-525 MG01-3280-525 MG013279525807 20003278525807 MG013278525807 20013280525807 20013279525807 20013278525807 20003280525807 20003279525807 MG013280525807 2002-3278-525 2002-3278-807 2002-3279-525 2002-3279-807 2002-3280-525 2002-3280-807 95616802328058 95616801327958 95616801328052 95616801328058 95616802327852 95616802327858 95616802327952 95616802328052 95616801327852 95616811327852 95616811327858 95616811327952 95616811327958 95616811328052 95616811328058 95616802327958 95616801327952 95616801327858 9561023279807 9561-2000-3277 9561-6801-3277 9561-MG01-3277 9561-2000-3278 9561-MG01-3278 9561-MG02-3280 9561-2000-3279 95612000327852 9561MG01327752 9561-MG01-3279 95616907327852 9561MG013277 95612000327980 9561MG01327980 9561E-MG01-327 9561-SP02-3278 9561-SP02-3277 9561-SP02-3279 9561-SP02-3280 9561-SP01-3280
132-1430EA 7970 DKK
132-1430 132-1431 132-1432 132-1433 132-1434 132-1435 132-1448 132-1449 132-1450 132-0905 132-0901 132-0904 BIMO20013280525807 BIMO20013279525807 132-1436 132-0903 132-0902 132-0906 132-1437 132-1438 132-1439 132-1440 132-1441 132-1442 BIMO95616802328058 132-0908 BIMO95616801328052 132-0909 BIMO95616802327852 BIMO95616802327858 BIMO95616802327952 BIMO95616802328052 BIMO95616801327852 BIMO95616811327852 BIMO95616811327858 BIMO95616811327952 BIMO95616811327958 BIMO95616811328052 BIMO95616811328058 BIMO95616802327958 BIMO95616801327952 132-0907 BIMO9561023279807 132-0976 132-0978 132-0977 139-0629 139-0630 BIMO9561-MG02-3280 BIMO9561-2000-3279 BIMO95612000327852 BIMO9561MG01327752 BIMO9561-MG01-3279 BIMO95616907327852 BIMO9561MG013277 BIMO95612000327980 BIMO9561MG01327980 BIMO9561E-MG01-327 BIMO9561-SP02-3278 BIMO9561-SP02-3277 BIMO9561-SP02-3279 BIMO9561-SP02-3280 BIMO9561-SP01-3280
Production and laboratory chairs, with permanent contact backrest, Neon 3
Stole
Neon supports the forward-leaning sitting position that is often necessary in lab work. Its modern design matches the high-tech laboratory environment perfectly and of course it has all the characteristics you would expect of a good laboratory chair. It is seamless, easy to clean, washable, and resistant to disinfectants. So, Neon is the ideal solution for the laboratory, when a really comfortable ergonomic chair is needed.
  • Permanent contact backrest
  • Ergonomic package with height, seat inclination, seat depth and backrest height adjustment plus weight regulation
  • Robust design for a long service life
  • Very few composite materials
  • Easy change of upholstery
  • Flex strip available in three colours
  • Five-star base made of aluminium; powder coated black
  • Anti-shock lock
  • Certified sustainable production
Permanent contact backrest: It follows the user's movements and ensures that the back always remains fully supported. Weight regulation ensures that the balance between the resistance of the backrest and the body weight is always steady. The permanent contact backrest is ideal for work where people have to lean forward for a long time, with only small distance between the eyes and workpiece (example: Working at close range in a laboratory).
Certificeringer: Blue Angel Eco Label; non toxic; complies with DIN 68 877 standards; Conforms to quality management system DIN EN ISO 9001 and complies with regulation EWG Nr. 1836/93 and Environmental Management System DIN EN ISO14001