VWR®, Sample Vials, with Snap-Caps
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-1758EA 344 DKK
216-1758 216-1759 216-1760 216-1770 216-1776 216-1777 216-1778 216-1782
VWR®, Sample Vials, with Snap-Caps
Hætteglas
Soda-lime glass, clear.
  • Ideal for storing medication as liquid, powder or capsules
  • Not suitable for heat applications
Leveringsinformation: Delivered in practical safe trays. PE snap-caps are included.