VWR®, Culture Tubes with Black PP Screw Cap
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212-0656EA 273 DKK
212-0656 212-0657 212-0658 212-0659
VWR®, Culture Tubes with Black PP Screw Cap
Rør Dyrkningsrør
Made from soda-lime glass, clear.
  • GL thread
  • Wall thickness 1 mm (±0,04 mm)
  • High chemical and mechanical resistance
Advarsel: Note: Caps are not autoclavable.