VWR®, Aluminium foil in dispensers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
291-0044EA 417 DKK
291-0044 291-0046 291-0045 291-0047
VWR®, Aluminium foil in dispensers
Aluminiumsfolie
Pure aluminium 99,0 to 99,5%.
  • Comes in a practical dispenser
  • Available in various widths and thicknesses
  • Roll length to suit various requirements
Ideal for packaging, covering and insulation.