Hydrometers for sodium chloride
Leverandør: Ludwig Schneider GmbH & CO. KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30025112 3002511
SCNL30025112EA 970 DKK
SCNL30025112 SCNL3002511
Hydrometers for sodium chloride
Flydevægte
Made of high quality plastics, these hydrometers are designed for measurements in salt solutions.
  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Bestillingsoplysninger: A works calibration certificate is available on request.
Emballering: In a square plastic sleeve.